Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-tert-Butyl 4-(hydroxymethyl)-2,2-dimethyloxazolidine-3-carboxylate 97%

Supplier: AMBEED, INC
Warning

A137929-1G A137929-100MG A137929-250MG A137929-1G A137929-5G
103802-530EA 52 USD InStock
103802-530 77109-658 77109-660 77298-936 77315-526
(R)-tert-Butyl 4-(hydroxymethyl)-2,2-dimethyloxazolidine-3-carboxylate 97%
(R)-tert-Butyl 4-(hydroxymethyl)-2,2-dimethyloxazolidine-3-carboxylate
Precaución: for Research Use Only


Formula: C₁₁H₂₁NO₄
Boiling Pt: 314.07 °C (760 mm Hg)
Flash Pt: 143.745 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06202686
CAS Number: 108149-63-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty