Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-(+)-3-Boc-2,2-dimethyloxazolidine-4-carboxaldehyde 97%

Supplier: AMBEED, INC
Warning

A193576-1G A193576-5G A193576-100MG A193576-250MG
103816-554EA 25 USD InStock
103816-554 103847-170 77111-882 77111-884
(R)-(+)-3-Boc-2,2-dimethyloxazolidine-4-carboxaldehyde 97%
(R)-(+)-3-Boc-2,2-dimethyloxazolidine-4-carboxaldehyde
Precaución: for Research Use Only


Formula: C₁₁H₁₉NO₄
MW: 229.276 g/mol
Flash Pt: 102 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00674064
CAS Number: 95715-87-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty