Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R,R)-(-)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminocobalt(II) 99%

Supplier: AMBEED, INC
Warning

A132162-1G A132162-10G A132162-25G A132162-1G A132162-100G A132162-500G
103801-696EA 14 USD InStock
103801-696 77136-932 77136-934 77314-252 77745-947 77745-948
(R,R)-(-)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminocobalt(II) 99%
(R,R)-(-)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminocobalt(II)
Precaución: for Research Use Only


Formula: C₃₆H₅₂CoN₂O₂
MW: 603.74 g/mol
Melting Pt: >350 °C
Storage Temperature: Ambient
MDL Number: MFCD01631277
CAS Number: 176763-62-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty