Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(-)-Perillaldehyde 90% (by GC)

Supplier: AMBEED, INC
Warning

A284750-10G A284750-100G A284750-1G
103834-784EA 15 USD InStock
103834-784 77593-752 77685-809 77685-810
(-)-Perillaldehyde 90% (by GC)
(-)-Perillaldehyde
Precaución: Inert atmosphere, store in freezer, under –20 °C.


Formula: C₁₀H₁₄O
Boiling Pt: 238 °C (760 mm Hg)
Flash Pt: 96 °C
Storage Temperature: Freezer
MDL Number: MFCD00001543
CAS Number: 18031-40-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty