Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-(1,4-Dioxan-2-yl)methanol 97%

Supplier: AMBEED, INC
Warning

A291653-1G A291653-1G A291653-5G A291653-100MG A291653-250MG A291653-25G A291653-50MG
103813-918EA 22 USD InStock
103813-918 103835-058 76837-654 76837-656 77166-812 77166-814 77166-816 77166-818
(S)-(1,4-Dioxan-2-yl)methanol 97%
(S)-(1,4-Dioxan-2-yl)methanol
Precaución: for Research Use Only


Formula: C₅H₁₀O₃
Boiling Pt: 208.2±15.0 °C (760 mm Hg)
Flash Pt: 96.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD16652371
CAS Number: 406913-93-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty