Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-2-Amino-1,1,1-trifluoropropane hydrochloride 95%

Supplier: AMBEED, INC
Warning

A196475-5G A196475-1G A196475-25G A196475-5G A196475-100G A196475-100MG A196475-10G A196475-250MG
103816-852EA 43 USD InStock
103816-852 103847-464 77085-418 77085-420 77085-422 77139-252 77139-254 77139-256 77139-258
(S)-2-Amino-1,1,1-trifluoropropane hydrochloride 95%
(S)-2-Amino-1,1,1-trifluoropropane hydrochloride
Precaución: for Research Use Only


Formula: C₃H₆F₃N·ClH
Storage Temperature: Refrigerator
MDL Number: MFCD07779094
CAS Number: 125353-44-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty