Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-(-)-1-Boc-3-aminopyrrolidine 97%

Supplier: AMBEED, INC
Danger

A139221-1G A139221-10G A139221-250MG A139221-1G A139221-25G A139221-5G
103802-712EA 6 USD InStock
103802-712 77272-406 77272-408 77315-788 77315-790 77315-792
(S)-(-)-1-Boc-3-aminopyrrolidine 97%
(S)-(-)-1-Boc-3-aminopyrrolidine
Precaución: for Research Use Only


Formula: C₉H₁₈N₂O₂
Boiling Pt: 216…217 °C (760 mm Hg)
Flash Pt: 91 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03419271
CAS Number: 147081-44-5
UN: 2810
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty