Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-3-Amino-1-methylpiperidine 95%

Supplier: AMBEED, INC
Danger

A202025-1G A202025-5G A202025-100MG A202025-1G A202025-5G
103817-348EA 240 USD InStock
103817-348 103847-984 76986-502 77086-074 77086-076
(S)-3-Amino-1-methylpiperidine 95%
(S)-3-Amino-1-methylpiperidine
Precaución: for Research Use Only


Formula: C₆H₁₄N₂
Boiling Pt: 136.5±8.0 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD11894699
CAS Number: 902152-76-5
UN: 2920
ADR: 8,3,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty