Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-L-histidine 98%

Supplier: AMBEED, INC
Warning

A104111-1G A104111-5G A104111-100MG A104111-10G A104111-250MG A104111-1G A104111-5G A104111-25G A104111-100G
103796-656EA 8 USD InStock
103796-656 103822-368 77148-354 77148-356 77148-358 77309-228 77309-230 77610-806 77684-946
Fmoc-L-histidine 98%
Fmoc-L-histidine
Precaución: for Research Use Only


Formula: C₂₁H₁₉N₃O₄
MW: 377.4 g/mol
Boiling Pt: 706.7±60.0 °C (760 mm Hg)
Flash Pt: 381.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00190885
CAS Number: 116611-64-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty