Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-(S)-2-amino-3-hydroxy-3-methylbutanoic acid 97%

Supplier: AMBEED, INC
Warning

A100018-1G A100018-250MG A100018-5G
103795-198EA 32 USD InStock
103795-198 77160-142 77548-850
Fmoc-(S)-2-amino-3-hydroxy-3-methylbutanoic acid 97%
Fmoc-(S)-2-amino-3-hydroxy-3-methylbutanoic acid
Precaución: for Research Use Only


Formula: C₂₀H₂₁NO₅
Storage Temperature: Refrigerator
MDL Number: MFCD04974410
CAS Number: 1217603-41-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty