Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-2-(((Benzyloxy)carbonyl)amino)-3-(2-fluorophenyl)propanoic acid 95%

Supplier: AMBEED, INC
Warning

A101362-1G A101362-100MG A101362-250MG A101362-1G A101362-5G
103795-932EA 66 USD InStock
103795-932 76687-036 76687-038 77297-068 77623-822
(S)-2-(((Benzyloxy)carbonyl)amino)-3-(2-fluorophenyl)propanoic acid 95%
(S)-2-(((Benzyloxy)carbonyl)amino)-3-(2-fluorophenyl)propanoic acid
Precaución: for Research Use Only


Formula: C₁₇H₁₆FNO₄
MW: 317.31 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00797567
CAS Number: 127862-88-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty