Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-2-(1,3-Dioxoisoindolin-2-yl)propanoic acid 97%

Supplier: AMBEED, INC
Warning

A152314-5G A152314-100G A152314-10G A152314-25G
103842-542EA 10 USD InStock
103842-542 77095-862 77095-864 77318-156
(S)-2-(1,3-Dioxoisoindolin-2-yl)propanoic acid 97%
(S)-2-(1,3-Dioxoisoindolin-2-yl)propanoic acid
Precaución: for Research Use Only


Formula: C₁₁H₉NO₄
Melting Pt: 139…141 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00190990
CAS Number: 4192-28-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty