Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2S)-2-(3,4,5-Trimethoxyphenyl)butanoic acid 97%

Supplier: AMBEED, INC
Warning

A151284-1G A151284-5G A151284-100MG A151284-10G A151284-250MG A151284-25G A151284-1G A151284-5G
103804-242EA 102 USD InStock
103804-242 103842-390 76719-742 76719-744 76719-746 76719-748 77317-934 77317-936
(2S)-2-(3,4,5-Trimethoxyphenyl)butanoic acid 97%
(2S)-2-(3,4,5-Trimethoxyphenyl)butanoic acid
Precaución: for Research Use Only


Formula: C₁₃H₁₈O₅
Boiling Pt: 378.9 °C (760 mm Hg)
Flash Pt: 138.7 °C
Storage Temperature: Ambient
MDL Number: MFCD06245370
CAS Number: 195202-08-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty