Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-2-(3-Fluorophenyl)pyrrolidine 95%

Supplier: AMBEED, INC
Warning

A166749-1G A166749-100MG A166749-250MG A166749-1G
103806-094EA 564 USD InStock
103806-094 77191-296 77191-298 77320-634
(S)-2-(3-Fluorophenyl)pyrrolidine 95%
(S)-2-(3-Fluorophenyl)pyrrolidine
Precaución: Keep in a dark place, sealed in dry, at 2…8°C


Formula: C₁₀H₁₂FN
Storage Temperature: Refrigerator
MDL Number: MFCD06762565
CAS Number: 920274-04-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty