Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid 98%

Supplier: AMBEED, INC
Warning

A296148-5G A296148-25G A296148-100G A296148-10G A296148-250MG A296148-500G A296148-5G
103835-224EA 9 USD InStock
103835-224 76837-888 77002-418 77002-420 77002-422 77002-424 77305-688
(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid 98%
(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid
Precaución: for Research Use Only


Formula: C₁₄H₁₄O₃
Boiling Pt: 403.8 °C (760 mm Hg)
Melting Pt: 152…154 °C
Storage Temperature: Ambient
MDL Number: MFCD00010500
CAS Number: 22204-53-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty