Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-2-Acetamido-2-phenylacetic acid 95%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A134992-1G A134992-100MG A134992-10G A134992-250MG A134992-1G A134992-5G A134992-25G
103802-078EA 22 USD InStock
103802-078 76983-940 76983-942 76983-944 77314-852 77314-854 77683-194
(S)-2-Acetamido-2-phenylacetic acid 95%, Ambeed.Inc
(S)-2-Acetamido-2-phenylacetic acid
Precaución: For research use only.


Formula: C₁₀H₁₁NO₃
MW: 193.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20638234
CAS Number: 42429-20-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty