Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-2-Amino-2-cyclohexylacetic acid hydrochloride 97%

Supplier: AMBEED, INC
Warning

A185902-25G A185902-5G A185902-100G A185902-10G
103846-466EA 6 USD InStock
103846-466 77084-110 77084-112 77164-046 77164-048
(S)-2-Amino-2-cyclohexylacetic acid hydrochloride 97%
(S)-2-Amino-2-cyclohexylacetic acid hydrochloride
Precaución: for Research Use Only


Formula: C₈H₁₆ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD00050539
CAS Number: 191611-20-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty