Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-2-Methylpropane-2-sulfinamide 98%

Supplier: AMBEED, INC
Warning

A114422-100G A114422-10G A114422-25G A114422-500G A114422-5G A114422-1000G
103824-238EA 5 USD InStock
103824-238 77284-374 77284-376 77284-378 77284-380 77311-052 77747-830
(S)-2-Methylpropane-2-sulfinamide 98%
(S)-2-Methylpropane-2-sulfinamide
Precaución: for Research Use Only


Formula: C₄H₁₁NOS
Boiling Pt: 220 °C (760 mm Hg)
Melting Pt: 97…101 °C
Flash Pt: 75.1 °C
Storage Temperature: Ambient
MDL Number: MFCD05861480
CAS Number: 343338-28-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty