Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-2-Methylpyrrolidine hydrochloride 98%

Supplier: AMBEED, INC
Warning

A120004-1G A120004-10G A120004-25G A120004-1G
103799-868EA 27 USD InStock
103799-868 77094-418 77094-420 77311-976
(S)-2-Methylpyrrolidine hydrochloride 98%
(S)-2-Methylpyrrolidine hydrochloride
Precaución: for Research Use Only


Formula: C₅H₁₂ClN
Boiling Pt: 146.9 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD06796611
CAS Number: 174500-74-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty