Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-3-((S)-5-(4-Fluorophenyl)-5-hydroxypentanoyl)-4-phenyloxazolidin-2-one 97%

Supplier: AMBEED, INC
Warning

A140910-1G A140910-10G A140910-25G A140910-1G A140910-5G
103802-920EA 6 USD InStock
103802-920 76871-210 76871-212 77299-172 77316-040
(S)-3-((S)-5-(4-Fluorophenyl)-5-hydroxypentanoyl)-4-phenyloxazolidin-2-one 97%
(S)-3-((S)-5-(4-Fluorophenyl)-5-hydroxypentanoyl)-4-phenyloxazolidin-2-one
Precaución: for Research Use Only


Formula: C₂₀H₂₀FNO₄
Storage Temperature: Refrigerator
MDL Number: MFCD13194798
CAS Number: 189028-95-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty