Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-β3-phenylalanine 97%

Supplier: AMBEED, INC
Warning

A103563-1G A103563-5G A103563-10G A103563-250MG A103563-25G A103563-1G A103563-5G
103796-572EA 6 USD InStock
103796-572 103822-278 77245-242 77245-244 77245-246 77309-154 77309-156
(S)-β3-phenylalanine 97%
(S)-β3-phenylalanine
Precaución: for Research Use Only


Formula: C₉H₁₁NO₂
Boiling Pt: 307.5 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD01076238
CAS Number: 40856-44-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty