Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-3-Methyl-2-phenylbutanoic acid 98%

Supplier: AMBEED, INC
Warning

A158207-1G A158207-100MG A158207-250MG
103805-106EA 187 USD InStock
103805-106 103843-314 77028-482 77028-484
(S)-3-Methyl-2-phenylbutanoic acid 98%
(S)-3-Methyl-2-phenylbutanoic acid
Precaución: for Research Use Only


Formula: C₁₁H₁₄O₂
Melting Pt: 86…88 °C
Storage Temperature: Ambient
MDL Number: MFCD00167435
CAS Number: 13490-69-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty