Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-Ethyl 2-amino-6-((tert-butoxycarbonyl)amino)hexanoate hydrochloride 95%

Supplier: AMBEED, INC
Warning

A102095-1G A102095-100MG A102095-250MG
103796-298EA 109 USD InStock
103796-298 103821-990 76869-186 76869-188
(S)-Ethyl 2-amino-6-((tert-butoxycarbonyl)amino)hexanoate hydrochloride 95%
(S)-Ethyl 2-amino-6-((tert-butoxycarbonyl)amino)hexanoate hydrochloride
Precaución: for Research Use Only


Formula: C₁₃H₂₇ClN₂O₄
Storage Temperature: Refrigerator
MDL Number: MFCD23140990
CAS Number: 122456-82-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty