Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-Isopropyl 2-amino-3-(4-hydroxyphenyl)propanoate 97%

Supplier: AMBEED, INC
Warning

A200999-1G A200999-5G A200999-1G A200999-25G A200999-5G A200999-10G
103817-258EA 10 USD InStock
103817-258 103847-894 77085-960 77085-962 77085-964 77192-306
(S)-Isopropyl 2-amino-3-(4-hydroxyphenyl)propanoate 97%
(S)-Isopropyl 2-amino-3-(4-hydroxyphenyl)propanoate
Precaución: for Research Use Only


Formula: C₁₂H₁₇NO₃
Boiling Pt: 350.2 °C (760 mm Hg)
Flash Pt: 165.6 °C
Storage Temperature: Freezer
MDL Number: MFCD00871218
CAS Number: 126173-94-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty