Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-(+)-Ketoprofen 98%

Supplier: AMBEED, INC
Danger

A305587-1G A305587-5G A305587-100MG A305587-250MG A305587-25MG A305587-1MG
103814-438EA 31 USD InStock
103814-438 103835-604 77194-518 77194-520 77194-522 77746-581
(S)-(+)-Ketoprofen 98%
(S)-(+)-Ketoprofen
Precaución: for Research Use Only


Formula: C₁₆H₁₄O₃
MW: 254.29 g/mol
Boiling Pt: 431.316 °C (760 mm Hg)
Melting Pt: 93…96 °C (760 mm Hg)
Flash Pt: 228.793 °C
Storage Temperature: Ambient
MDL Number: MFCD00673316
CAS Number: 22161-81-5
UN: 2811
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty