Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-Methyl 2-(((benzyloxy)carbonyl)amino)but-3-enoate 96%

Supplier: AMBEED, INC
Warning

A114823-1G A114823-100MG A114823-1G
103798-484EA 372 USD InStock
103798-484 103824-326 77189-298 77297-336
(S)-Methyl 2-(((benzyloxy)carbonyl)amino)but-3-enoate 96%
(S)-Methyl 2-(((benzyloxy)carbonyl)amino)but-3-enoate
Precaución: for Research Use Only


Formula: C₁₃H₁₅NO₄
MW: 249.26 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00145552
CAS Number: 75266-40-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty