Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-Methyl-2-(pyrrolidin-2-yl)acetate hydrochloride 95%

Supplier: AMBEED, INC
Warning

A193295-1G A193295-100MG A193295-250MG A193295-1G
103816-520EA 279 USD InStock
103816-520 103847-134 76873-126 76873-128 77302-334
(S)-Methyl-2-(pyrrolidin-2-yl)acetate hydrochloride 95%
(S)-Methyl-2-(pyrrolidin-2-yl)acetate hydrochloride
Precaución: for Research Use Only


Formula: C₇H₁₄ClNO₂
Melting Pt: 53 °C
Storage Temperature: Refrigerator
MDL Number: MFCD22200809
CAS Number: 259868-83-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty