Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-Methyl-2-hydroxy-3-phenylpropanoate 98%

Supplier: AMBEED, INC
Warning

A166893-5G A166893-100G A166893-10G A166893-25G A166893-5G
103844-374EA 9 USD InStock
103844-374 76690-076 76690-078 77300-858 77320-658
(S)-Methyl-2-hydroxy-3-phenylpropanoate 98%
(S)-Methyl-2-hydroxy-3-phenylpropanoate
Precaución: for Research Use Only


Formula: C₁₀H₁₂O₃
MW: 180.2005 g/mol
Boiling Pt: 286.1 °C (760 mm Hg)
Melting Pt: 40…45 °C
Storage Temperature: Ambient
MDL Number: MFCD00038224
CAS Number: 13673-95-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty