Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-2-Octyl 4-[4-(Hexyloxy)benzoyloxy]benzoate 95%

Supplier: AMBEED, INC
Warning

A316752-1G A316752-250MG A316752-1G A316752-5G
103825-116EA 26 USD InStock
103825-116 103836-046 76723-904 76838-884 76838-886
(S)-2-Octyl 4-[4-(Hexyloxy)benzoyloxy]benzoate 95%
(S)-2-Octyl 4-[4-(Hexyloxy)benzoyloxy]benzoate
Precaución: for Research Use Only


Formula: C₂₈H₃₈O₅
MW: 454.61 g/mol
Boiling Pt: 565.8 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD01941070
CAS Number: 87321-20-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty