Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-tert-Butyl 2-methylpyrrolidine-1-carboxylate 97%

Supplier: AMBEED, INC
Danger

A257390-1G A257390-5G A257390-10G A257390-250MG A257390-1G A257390-5G
103812-300EA 40 USD InStock
103812-300 103852-168 77193-740 77193-742 77304-698 77304-700
(S)-tert-Butyl 2-methylpyrrolidine-1-carboxylate 97%
(S)-tert-Butyl 2-methylpyrrolidine-1-carboxylate
Precaución: for Research Use Only


Formula: C₁₀H₁₉NO₂
MW: 185.26 g/mol
Boiling Pt: 234 °C (760 mm Hg)
Flash Pt: 82.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06858469
CAS Number: 137496-71-0
UN: 2922
ADR: 8,6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty