Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-tert-Butyl azepan-3-ylcarbamate 97%

Supplier: AMBEED, INC
Warning

A113261-1G A113261-100MG A113261-250MG
103798-204EA 448 USD InStock
103798-204 103824-042 77025-862 77025-864
(S)-tert-Butyl azepan-3-ylcarbamate 97%
(S)-tert-Butyl azepan-3-ylcarbamate
Precaución: for Research Use Only


Formula: C₁₁H₂₂N₂O₂
MW: 214.31 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18831516
CAS Number: 213990-48-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty