Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3S)-3-Amino-N-methylpyrrolidine, N-BOC protected 97%

Supplier: AMBEED, INC
Warning

A145018-1G A145018-5G A145018-100MG A145018-250MG A145018-10G A145018-25G
103803-482EA 22 USD InStock
103803-482 103841-562 76972-786 76972-788 77881-131 77881-132
(3S)-3-Amino-N-methylpyrrolidine, N-BOC protected 97%
(3S)-3-Amino-N-methylpyrrolidine, N-BOC protected
Precaución: for Research Use Only


Formula: C₁₀H₂₀N₂O₂
MW: 200.278 g/mol
Boiling Pt: 268.3 °C (760 mm Hg)
Flash Pt: 116 °C
Storage Temperature: Ambient
MDL Number: MFCD08703388
CAS Number: 169750-01-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty