Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Phenyl vinyl sulphone 97%

Supplier: AMBEED, INC
Danger

A141179-1G A141179-25G A141179-1G A141179-5G
103802-962EA 9 USD InStock
103802-962 77246-946 77299-206 77316-110
Phenyl vinyl sulphone 97%
Phenyl vinyl sulphone
Precaución: for Research Use Only


Formula: C₈H₈O₂S
MW: 168.22 g/mol
Boiling Pt: 314.4 °C (760 mm Hg)
Melting Pt: 67…69 °C
Flash Pt: 178.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00007554
CAS Number: 5535-48-8
UN: 1759
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty