Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

cis-2-Butene-1,4-diol 96%

Supplier: AMBEED, INC
Warning

A1001233-100G A1001233-25G A1001233-500G
103820-982EA 5 USD InStock
103820-982 76645-868 76645-870 76645-872 76645-874
cis-2-Butene-1,4-diol 96%
cis-2-Butene-1,4-diol
Precaución: for Research Use Only


Formula: C₄H₈O₂
MW: 88.11 g/mol
Boiling Pt: 235
Flash Pt: 128 °C
Storage Temperature: Ambient
MDL Number: MFCD00002924
CAS Number: 6117-80-2
Merck Index: 12,02390
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty