Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(Z)-3-Iodopropenoic acid methyl ester 95/5 Z/E 97%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A118602-1G A118602-1G A118602-250MG
103799-082EA 23 USD InStock
103799-082 77297-614 77683-068
(Z)-3-Iodopropenoic acid methyl ester 95/5 Z/E 97%, Ambeed.Inc
(Z)-3-Iodopropenoic acid methyl ester 95/5 Z/E
Precaución: For research use only.


Formula: C₄H₅IO₂
Boiling Pt: 182.1 °C (760 mm Hg)
Flash Pt: 63.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09265518
CAS Number: 6214-23-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty