Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

[1,2,4]Triazolo[4,3-a]pyridine-6-carboxylic acid 97%

Supplier: AMBEED, INC
Warning

A123271-1G A123271-100MG A123271-250MG
103800-378EA 73 USD InStock
103800-378 76766-716 77876-891
[1,2,4]Triazolo[4,3-a]pyridine-6-carboxylic acid 97%
[1,2,4]Triazolo[4,3-a]pyridine-6-carboxylic acid
Precaución: for Research Use Only


Formula: C₇H₅N₃O₂
Storage Temperature: Refrigerator
MDL Number: MFCD07376943
CAS Number: 933708-92-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty