Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

[2,2':6',2''-Terpyridine]-4,4',4''-tricarboxylic acid 95%

Supplier: AMBEED, INC
Warning

A114832-1G A114832-100MG A114832-250MG A114832-1G
103798-486EA 161 USD InStock
103798-486 103824-328 76822-058 76822-060 77311-150
[2,2':6',2''-Terpyridine]-4,4',4''-tricarboxylic acid 95%
[2,2':6',2''-Terpyridine]-4,4',4''-tricarboxylic acid
Precaución: for Research Use Only


Formula: C₁₈H₁₁N₃O₆
MW: 365.3 g/mol
Boiling Pt: 890.3 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD06796963
CAS Number: 216018-58-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty