Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

[2,2'-Bipyridine]-5,5'-diamine 97%

Supplier: AMBEED, INC
Warning

A353356-1G A353356-100MG A353356-250MG A353356-1G A353356-5G
103826-092EA 128 USD InStock
103826-092 77263-890 77263-892 77306-640 77748-697
[2,2'-Bipyridine]-5,5'-diamine 97%
[2,2'-Bipyridine]-5,5'-diamine
Precaución: for Research Use Only


Formula: C₁₀H₁₀N₄
Boiling Pt: 432.838 °C (760 mm Hg)
Flash Pt: 245.502 °C
Storage Temperature: Refrigerator
MDL Number: MFCD14636205
CAS Number: 52382-48-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty