Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-((Isocyano(p-tolyl)methyl)sulfonyl)-4-methylbenzene 95%

Supplier: AMBEED, INC
Warning

A102237-1G A102237-5G A102237-250MG A102237-1G
103796-342EA 49 USD InStock
103796-342 103822-034 77245-172 77245-174 77282-918
1-((Isocyano(p-tolyl)methyl)sulfonyl)-4-methylbenzene 95%
1-((Isocyano(p-tolyl)methyl)sulfonyl)-4-methylbenzene
Precaución: for Research Use Only


Formula: C₁₆H₁₅NO₂S
MW: 285.36 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04114796
CAS Number: 1330529-37-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty