Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(2-Chloro-4-(trifluoromethyl)phenyl)ethanone 98%

Supplier: AMBEED, INC
Warning

A169815-1G A169815-5G A169815-10G A169815-250MG A169815-1G
103806-414EA 33 USD InStock
103806-414 103844-678 76872-352 76872-354 77301-000
1-(2-Chloro-4-(trifluoromethyl)phenyl)ethanone 98%
1-(2-Chloro-4-(trifluoromethyl)phenyl)ethanone
Precaución: for Research Use Only


Formula: C₉H₆ClF₃O
Storage Temperature: Ambient
MDL Number: MFCD13194254
CAS Number: 129322-81-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty