Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2'-Hydroxy-5'-methylacetophenone 98%

Supplier: AMBEED, INC
Warning

A325803-25G A325803-5G A325803-100G A325803-10G A325803-1G A325803-500G
103836-360EA 18 USD InStock
103836-360 76839-306 76839-308 77002-938 77002-940 77002-942 77672-676
2'-Hydroxy-5'-methylacetophenone 98%
2'-Hydroxy-5'-methylacetophenone
Precaución: for Research Use Only


Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Melting Pt: 53
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00002380
CAS Number: 1450-72-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty