Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(3,4-Dichlorophenyl)urea 95%

Supplier: AMBEED, INC
Warning

A132348-1G A132348-10G A132348-250MG A132348-1G A132348-5G
103801-726EA 79 USD InStock
103801-726 77190-108 77190-110 77298-554 77298-556
1-(3,4-Dichlorophenyl)urea 95%
1-(3,4-Dichlorophenyl)urea
Precaución: for Research Use Only


Formula: C₇H₆Cl₂N₂O
Storage Temperature: Refrigerator
MDL Number: MFCD00025417
CAS Number: 2327-02-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty