Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3'-Bromo-2'-hydroxyacetophenone 97%

Supplier: AMBEED, INC
Warning

A170792-1G A170792-5G A170792-1G A170792-25G A170792-5G A170792-250MG
103806-542EA 29 USD InStock
103806-542 103844-818 77081-932 77081-934 77081-936 77191-398
3'-Bromo-2'-hydroxyacetophenone 97%
3'-Bromo-2'-hydroxyacetophenone
Precaución: for Research Use Only


Formula: C₈H₇BrO₂
Storage Temperature: Ambient
MDL Number: MFCD08236725
CAS Number: 1836-05-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty