Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoro-N-methylbenzylamine 97%

Supplier: AMBEED, INC
Danger

A145548-1G A145548-5G A145548-100G A145548-10G A145548-5G A145548-1G A145548-25G
103803-552EA 7 USD InStock
103803-552 103841-636 76811-970 76811-972 77299-462 77316-940 77316-942
3-Fluoro-N-methylbenzylamine 97%
3-Fluoro-N-methylbenzylamine
Precaución: for Research Use Only


Formula: C₈H₁₀FN
MW: 139.17 g/mol
Boiling Pt: 183…184 °C (760 mm Hg)
Flash Pt: 66 °C
Storage Temperature: Ambient
MDL Number: MFCD04623547
CAS Number: 90389-84-7
UN: 2735
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty