Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)ethanone 98%

Supplier: AMBEED, INC
Warning

A116554-1G A116554-100MG A116554-250MG
103798-754EA 200 USD InStock
103798-754 103824-618 77245-942 77745-860
1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)ethanone 98%
1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)ethanone
Precaución: for Research Use Only


Formula: C₁₃H₁₈BNO₃
Boiling Pt: 369.8 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09952047
CAS Number: 741709-58-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty