Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(4-(Benzyloxy)-3-nitrophenyl)-2-bromoethanone 97%

Supplier: AMBEED, INC
Warning

A279163-1G A279163-5G A279163-1G A279163-5G A279163-10G A279163-250MG
103813-456EA 8 USD InStock
103813-456 103853-400 77093-332 77093-334 77250-482 77250-484
1-(4-(Benzyloxy)-3-nitrophenyl)-2-bromoethanone 97%
1-(4-(Benzyloxy)-3-nitrophenyl)-2-bromoethanone
Precaución: for Research Use Only


Formula: C₁₅H₁₂BrNO₄
Boiling Pt: 465.2 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD06797599
CAS Number: 43229-01-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty