Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(4-Bromo-2-nitrophenyl)ethanone 98%

Supplier: AMBEED, INC
Warning

A164052-1G A164052-5G A164052-250MG A164052-25G
103805-804EA 25 USD InStock
103805-804 103844-070 76973-696 76973-698
1-(4-Bromo-2-nitrophenyl)ethanone 98%
1-(4-Bromo-2-nitrophenyl)ethanone
Precaución: for Research Use Only


Formula: C₈H₆BrNO₃
MW: 244.04 g/mol
Boiling Pt: 345.9 °C (760 mm Hg)
Melting Pt: 70 °C
Storage Temperature: Ambient
MDL Number: MFCD11977406
CAS Number: 90004-94-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty