Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(4-Bromobenzyl)-1H-imidazole 95%

Supplier: AMBEED, INC
Warning

A124549-1G A124549-250MG A124549-1G A124549-5G
103800-548EA 16 USD InStock
103800-548 77189-830 77189-832 77298-000 77312-686
1-(4-Bromobenzyl)-1H-imidazole 95%
1-(4-Bromobenzyl)-1H-imidazole
Precaución: for Research Use Only


Formula: C₁₀H₉BrN₂
Boiling Pt: 371.5 °C (760 mm Hg)
Melting Pt: 75 °C
Storage Temperature: Ambient
MDL Number: MFCD06421442
CAS Number: 72459-46-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty