Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(4-Bromophenyl)-N-methylmethanamine 97%

Supplier: AMBEED, INC
Warning

A365209-1G A365209-25G A365209-5G A365209-100G A365209-10G
103826-376EA 11 USD InStock
103826-376 103837-396 76840-630 76840-632 76840-634 77115-344 77115-346
1-(4-Bromophenyl)-N-methylmethanamine 97%
1-(4-Bromophenyl)-N-methylmethanamine
Precaución: for Research Use Only


Formula: C₈H₁₀BrN
Boiling Pt: 236.3 °C (760 mm Hg)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD04632143
CAS Number: 699-03-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty