Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(4-Chlorobutyl)-4-fluorobenzene 95%

Supplier: AMBEED, INC
Warning

A102077-1G A102077-250MG A102077-1G
103796-282EA 89 USD InStock
103796-282 76869-184 77308-864
1-(4-Chlorobutyl)-4-fluorobenzene 95%
1-(4-Chlorobutyl)-4-fluorobenzene
Precaución: for Research Use Only


Formula: C₁₀H₁₂ClF
MW: 186.66
Boiling Pt: 253.5 °C (760 mm Hg)
Flash Pt: 120.2 °C
Storage Temperature: Ambient
MDL Number: MFCD11935101
CAS Number: 54540-58-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty